Skip to Content
Merck
All Photos(1)

Key Documents

SAB4200132

Sigma-Aldrich

Anti-ULK3 antibody produced in rabbit

~1 mg/mL, affinity isolated antibody

Synonym(s):

Anti-unc-51-like kinase 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.44

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~50 kDa

species reactivity

human

packaging

antibody small pack of 25 μL

concentration

~1 mg/mL

technique(s)

western blot: 1-2 μg/mL using whole extracts of human U87 cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ULK3(25989)

General description

Anti-ULK3 is produced in rabbit using as the immunogen a synthetic peptide corresponding to a fragment of human ULK3, conjugated to KLH. Unc-51-like kinase 3 (ULK3) belongs to the ULK family of serine/threonine kinases. ULK3 is an autophagy related protein with an N-terminal kinase domain, which shows high similarity to those of ULK1 and ULK2. ULK3 localizes to the elongating isolation membrane upon autophagy activation.

Application

Anti-ULK3 antibody produced in rabbit has been used in immunoblotting.

Biochem/physiol Actions

Unc-51-like kinase 3 (ULK3) controls abscission timing in response to lagging chromosomes. Overexpression of ULK3 induces autophagy and senescence. ULK3 is also a positive regulator of the Hedgehog signalling pathway.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

mTOR regulation of autophagy
Jung CH, et al.
Febs Letters, 584(7), 1287-1295 (2010)
The ULK1 complex: sensing nutrient signals for autophagy activation
Wong PM, et al.
Autophagy, 9(2), 124-137 (2013)
Pharmacological activation of REV-ERBs is lethal in cancer and oncogene-induced senescence
Sulli G, et al.
Nature, 553(7688), 351-351 (2018)
Anna Caballe et al.
eLife, 4, e06547-e06547 (2015-05-27)
The endosomal sorting complexes required for transport (ESCRT) machinery mediates the physical separation between daughter cells during cytokinetic abscission. This process is regulated by the abscission checkpoint, a genome protection mechanism that relies on Aurora B and the ESCRT-III subunit
The ULK3 Kinase Is Critical for Convergent Control of Cancer-Associated Fibroblast Activation by CSL and GLI
Goruppi , et al.
Testing, 20(10), 2468-2479 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service