Skip to Content
Merck
All Photos(1)

Key Documents

SAB4200108

Sigma-Aldrich

Anti-TIN2 antibody, Mouse monoclonal

clone TIN30, purified from hybridoma cell culture

Synonym(s):

Anti-TERF1 (TRF1)-interacting nuclear factor 2, Anti-TIN2L, Anti-TINF2

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
$699.00

$699.00


Estimated to ship on23 April 2025


Request a Bulk Order

Select a Size

Change View
200 μL
$699.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$699.00


Estimated to ship on23 April 2025


Request a Bulk Order

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

TIN30, monoclonal

form

buffered aqueous solution

mol wt

antigen ~40 kDa

species reactivity

human

packaging

antibody small pack of 25 μL

concentration

~1.0 mg/mL

technique(s)

western blot: 0.5-1.0 μg/mL using HeLa cell extract

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TINF2(26277)

General description

Monoclonal Anti-TIN2 (mouse IgG1 isotype) is derived from the hybridoma TIN-30 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with a synthetic peptide corresponding to a fragment of human TIN-2, conjugated to keyhole limpet hemocyanin (KLH). The six shelterin subunits are: TRF1, TRF2, and POT1 which directly recognize the TTAGGG repeats, and TIN2, Rap1 and TPP1. TIN2 (TRF-1 interacting nuclear factor 2) is recruited to the telomere through the TRF homology (TRFH) domain of TRF1 and tethers TPP1/POT1 to TRF1 and TRF2.

Application

Monoclonal Anti-TIN2 antibody produced in mouse has been used in immunoblotting.

Biochem/physiol Actions

TIN2 (TRF-1 interacting nuclear factor 2) also connects TRF1 to TRF2 and this link contributes to the stabilization of TRF2 on telomeres. In addition, TIN2 has also been found to tether the telomeres to the nuclear matrix. Interestingly, this protein has also been found to be mutated in Dyskeratosis Congenita, a multi-system disorder which in its classical form is characterized by abnormalities of the skin, nails, and mucous membranes.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A novel form of the telomere-associated protein TIN2 localizes to the nuclear matrix
Kaminker PG, et al.
Cell Cycle, 8(6), 931-939 (2009)
TIN2 Binds TRF1 and TRF2 Simultaneously and Stabilizes the TRF2 Complex on Telomeres
Ye JZS, et al.
The Journal of Biological Chemistry, 279(45), 47264-47271 (2004)
Titia de Lange
Genes & development, 19(18), 2100-2110 (2005-09-17)
Added by telomerase, arrays of TTAGGG repeats specify the ends of human chromosomes. A complex formed by six telomere-specific proteins associates with this sequence and protects chromosome ends. By analogy to other chromosomal protein complexes such as condensin and cohesin
David Frescas et al.
Genes & development, 28(2), 153-166 (2014-01-23)
The progressive bone marrow failure syndrome dyskeratosis congenita (DC) is often caused by mutations in telomerase or the factors involved in telomerase biogenesis and trafficking. However, a subset of DC patients is heterozygous for mutations in the shelterin component TIN2.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service