Skip to Content
Merck
All Photos(2)

Key Documents

SAB4200081

Sigma-Aldrich

Monoclonal Anti-BOP1 antibody produced in rat

~1.0 mg/mL, clone BOP 6H11, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-Block of proliferation 1

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
$694.00

$694.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
200 μL
$694.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$694.00


Estimated to ship on30 May 2025


Request a Bulk Order

biological source

rat

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

BOP 6H11, monoclonal

form

buffered aqueous solution

mol wt

antigen ~100 kDa

species reactivity

human

packaging

antibody small pack of 25 μL

concentration

~1.0 mg/mL

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: 0.5-1.0 μg/mL using HeLa cell extracts

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BOP1(23246)

General description

Monoclonal Anti- BOP1 (rat IgG1 isotype) is derived from the hybridoma BOP 6H11 produced by the fusion of mouse myeloma cells (P3X63Ag8.653) and splenocytes from rat immunized with a peptide corresponding to a fragment of human BOP1, coupled to ovalbumin.

Application

Monoclonal Anti-BOP1 antibody produced in rat has been used in several immunochemical techniques including immunoblotting, immunoprecipitation, and immunocytochemistry.

Biochem/physiol Actions

Ribosome biogenesis protein BOP1 is an essential component of A nucleolar complex, PeBoW that is involved in ribosome biogenesis and cell proliferation. Fluctuations in Bop1 protein level were found to affect the assembly and integrity of the whole PeBoW (Pes1, Bop1, and WDR12) complex. Overexpression of Bop1 mRNA is associated with the development of colorectal cancer.

Physical form

Solution in 0.01M phosphate buffered saline pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Interdependence of Pes1, Bop1, and WDR12 controls nucleolar localization and assembly of the PeBoW complex required for maturation of the 60S ribosomal subunit
Rohrmoser M, et al.
Molecular and Cellular Biology, 27(10), 3682-3694 (2007)
Siqi Li et al.
The Journal of pathology, 254(3), 265-278 (2021-04-03)
Chemoresistance is a major obstacle to the treatment of triple-negative breast cancer (TNBC), which has a poor prognosis. Increasing evidence has demonstrated the essential role of cancer stem cells (CSCs) in the process of TNBC chemoresistance. However, the underlying mechanism
Michael Hölzel et al.
The Journal of cell biology, 170(3), 367-378 (2005-07-27)
Target genes of the protooncogene c-myc are implicated in cell cycle and growth control, yet the linkage of both is still unexplored. Here, we show that the products of the nucleolar target genes Pes1 and Bop1 form a stable complex

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service