Skip to Content
Merck
All Photos(2)

Key Documents

SAB4200061

Sigma-Aldrich

Anti-CYLD (C-terminal) antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody

Synonym(s):

Anti-Cylindromatosis (turban tumor syndrome), deubiquitinating enzyme CYLD, Anti-Ubiquitin carboxyl-terminal hydrolase CYLD, Anti-Ubiquitin specific peptidase like 2, Anti-Ubiquitin thiolesterase CYLD

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
$822.00

$822.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
200 μL
$822.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$822.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~110 kDa

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

immunohistochemistry: 10-20 μg/mL using biotin / ExtrAvidin®-Peroxidase staining of heat-retrieved formalin-fixed, paraffin-embedded human skin sections
western blot: 2.5-5.0 μg/mL using whole extract of HEK-293T cells overexpressing human CYLD

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CYLD(1540)

Related Categories

General description

CYLD is a tumor suppressor protein exhibiting deubiquitination enzyme (DUB) activity, that specifically cleaves Lys63 -linked polyUb. CYLD contains a ubiquitin C-terminal hydrolase (UCH) domain, which is responsible for the removal of ubiquitin chains, and three cytoskeleton-associated protein-glycine conserved (CAP-Gly) domains, which are found in various microtubule-binding proteins.

Application

Anti-CYLD (C-terminal) antibody produced in rabbit has been used immunoblotting and immunohistochemistry.

Biochem/physiol Actions

CYLD physically interacts with many different proteins, some of which positively mediate signaling through the NF-κB and c-Jun N-terminal kinase (JNK) pathways and induces their deubiquitination. It controls cell proliferation, cell survival and inflammatory responses by negatively regulating NF-κB and/or JNK-signaling pathways. CYLD also regulates other physiological pathways such as cell cycle progression, spermatogenesis, and osteoclastogenesis. Mutations in the CYLD gene have been associated with cylindromatosis, multiple familial trichoepithelioma, and Brooke-Spiegler syndrome.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Legal Information

ExtrAvidin is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mutations in the CYLD gene in Brooke-Spiegler syndrome, familial cylindromatosis, and multiple familial trichoepithelioma: lack of genotype-phenotype correlation
Bowen S, et al.
The Journal of Investigative Dermatology, 124(5), 919-920 (2005)
Premature aging and cancer development in transgenic mice lacking functional CYLD
Alameda JP, et al.
Aging (Albany. NY.), 11(1), 127-127 (2019)
Mehdi Baratchian et al.
The Journal of biological chemistry, 291(14), 7608-7620 (2016-02-13)
The viral FLICE-like inhibitory protein (FLIP) protein from Kaposi sarcoma-associated herpesvirus activates the NF-κB pathway by forming a stable complex with a central region (amino acids 150-272) of the inhibitor of NF-κB kinase (IKK) γ subunits, thereby activating IKK. Cellular
Josefa P Alameda et al.
International journal of molecular sciences, 22(13) (2021-07-03)
Cylindromatosis (CYLD) is a deubiquitinase (DUB) enzyme that was initially characterized as a tumor suppressor of adnexal skin tumors in patients with CYLD syndrome. Later, it was also shown that the expression of functionally inactive mutated forms of CYLD promoted
The tumour suppressor CYLD negatively regulates NF-kappaB signalling by deubiquitination
Kovalenko A, et al.
Nature, 424(6950), 801-801 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service