Skip to Content
Merck
All Photos(3)

Key Documents

SAB4100383

Sigma-Aldrich

Monoclonal Anti-TMOD1 antibody produced in mouse

clone TMOD1-A328, culture supernatant

Synonym(s):

Anti-D9S57E, Anti-ETMOD, Anti-TMOD, Anti-tropomodulin 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

TMOD1-A328, monoclonal

form

aqueous solution

mol wt

antigen predicted mol wt 40.4 kDa

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG2b

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... TMOD1(7111)

General description

Tropomodulin 1 (TMOD1) is an actin-capping protein, which is expressed in the cardiac muscle and in slow-twitch muscle fibers. It possesses two actin- and two tropomyosin (TM)-binding regions. The gene encoding this protein is localized on human chromosome 9q.

Immunogen

TMOD1 purified recombinant protein

Biochem/physiol Actions

Tropomodulin 1 (TMOD1) prevents the elongation and depolymerization of actin filaments by binding to the slowly growing pointed end of the actin filament. It has been associated with triple-negative breast cancers (TNBC). TMOD1 may act as a stabilizer of β-catenin.

Physical form

Solution in Dulbecco′s Modified Eagle′s Medium with 10% fetal calf serum and 15 mM sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tropomodulin 1 Expression Driven by NF-?B Enhances Breast Cancer Growth
Taku Ito-Kureha
Cancer Research, 75(1) (2015)
Leucine 135 of Tropomodulin-1 Regulates Its Association with Tropomyosin, Its Cellular Localization, and the Integrity of Sarcomeres*
Kimi Y and Larry Kedes
The Journal of Biological Chemistry (2006)
Complete trisomy 9 with unusual phenotypic associations: Dandy-Walker malformation, cleft lip and cleft palate, cardiovascular abnormalities
Gabriele Tonni, et.al
Taiwanese Journal of Obstetrics & Gynecology, 53(4), 592-597 (2014)
Mechanism of actin filament pointed end capping by tropomodulin
Jampani Nageswara Rao
Science, 345(6195), 463-467 (2014)
Complete trisomy 9 with unusual phenotypic associations: Dandy-Walker malformation, cleft lip and cleft palate, cardiovascular abnormalities
GabrieleTonni
Taiwanese Journal of Obstetrics & Gynecology, 53(4), 592-597 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service