Skip to Content
Merck
All Photos(1)

Key Documents

SAB3701044

Sigma-Aldrich

Anti-Mouse IgG IgA IgM (H+L), highly cross adsorbed antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$301.00

$301.00


Check Cart for Availability


Select a Size

Change View
1 MG
$301.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

$301.00


Check Cart for Availability

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse

concentration

1.0 mg/mL

technique(s)

immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Specificity

Secondary Antibody was prepared from polyspecific antiserum by immunoaffinity chromatography using antigens coupled to agarose beads followed by cross adsorption to remove unwanted specificities. Assay by immunoelectrophoresis resulted in a single precipitin arc against Anti-Goat Serum, Mouse IgG, Mouse IgA and Mouse IgM. No reaction was observed against Bovine, Horse or Human Serum Proteins. This reagent is suitable for the detection of all Mouse isotypes and chain combinations. ELISA was used to confirm specificity at less than 1% of the target signal.

Immunogen

Mouse IgG IgA and IgM whole molecule

Physical properties

Antibody format: IgG

Physical form

Supplied in 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chao Lv et al.
Oncology letters, 15(1), 1103-1112 (2018-02-10)
Intraoperative manipulation causes circulating tumor cell (CTC) shedding into the blood and accelerates metastasis in non-small cell lung cancer (NSCLC). The present study was conducted to assess the degree of dissemination resulting from surgery and to explore the biological features
Yulan Zhen et al.
Oncology letters, 15(5), 6562-6570 (2018-05-05)
The effects of hydrogen sulfide (H2S) on cancer are controversial. Our group previously demonstrated that exogenous H2S promotes the development of cancer via amplifying the activation of the nuclear factor-κB signaling pathway in hepatocellular carcinoma (HCC) cells (PLC/PRF/5). The present
Yin-Ling Zhang et al.
Oncotarget, 12(4), 366-378 (2021-03-05)
The effects and mechanisms of folic acid (FA) as a chemopreventive agent for tumorigenesis of hepatocellular carcinoma (HCC) remain unclear. In this study, the QSG-7701, a human normal liver cell line, was cultured in different FA levels (High, Normal or
Ming Tu et al.
Molecular medicine reports, 15(4), 1713-1721 (2017-03-06)
Cluster of differentiation 164 (CD164), a sialomucin, has been demonstrated to be involved in the regulation of proliferation, apoptosis, adhesion and differentiation in multiple cancers. CD164 is regarded to be a potential promotor of tumor growth. However, the involvement of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service