Skip to Content
Merck
All Photos(1)

Key Documents

SAB3700690

Sigma-Aldrich

Anti-Sheep IgG F(ab′)2-Alkaline Phosphatase antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

ALP, Alk Phos

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$266.00

$266.00


Estimated to ship on28 May 2025



Select a Size

Change View
1 MG
$266.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

$266.00


Estimated to ship on28 May 2025


biological source

rabbit

conjugate

alkaline phosphatase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

sheep

concentration

0.6 mg/mL

technique(s)

immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Specificity

This product was prepared from monospecific antiserum by immunoaffinity chromatography using Sheep IgG coupled to agarose beads followed by solid phase adsorption(s) to remove any unwanted reactivities. Assay by immunoelectrophoresis resulted in a single precipitin arc against Anti-Alkaline Phosphatase (calf intestine), Anti-Rabbit Serum, Sheep IgG, Sheep IgG F(ab′)2 and Sheep Serum. No reaction was observed against Sheep IgG F(c).

Immunogen

Sheep IgG F(ab′)2 fragment

Physical properties

Antibody format: IgG

Physical form

Supplied in 0.05 M Tris Chloride, 0.15M Sodium Chloride, 0.001M Magnesium Chloride, 0.0001M Zinc Chloride, 50% (v/v) Glycerol; pH 8.0 with 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service