Skip to Content
Merck
All Photos(1)

Key Documents

SAB3700488

Sigma-Aldrich

Anti-HAMSTER IgG (H+L) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing

Select a Size

2 MG
$374.00

$374.00


Check Cart for Availability


Select a Size

Change View
2 MG
$374.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

$374.00


Check Cart for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

hamster

concentration

2.0 mg/mL

technique(s)

immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Related Categories

Specificity

This product was prepared from monospecific antiserum by immunoaffinity chromatography using Hamster IgG coupled to agarose beads. Assay by immunoelectrophoresis resulted in a single precipitin arc against Anti-Rabbit Serum, Golden Syrian Hamster IgG and Golden Syrian Hamster Serum. Greatly reduced reactivity will occur against Armenian Hamster IgG.

Immunogen

Golden Syrian Hamster IgG whole molecule

Physical properties

Antibody format: IgG

Physical form

Supplied in 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nguyen T Van et al.
Nature communications, 14(1), 7363-7363 (2023-11-15)
Environmental factors are the major contributor to the onset of immunological disorders such as ulcerative colitis. However, their identities remain unclear. Here, we discover that the amount of consumed L-Tryptophan (L-Trp), a ubiquitous dietary component, determines the transcription level of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service