Skip to Content
Merck
All Photos(1)

Key Documents

SAB3501203

Sigma-Aldrich

Anti-Avian Influenza Neuraminidase antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$753.00

$753.00


Check Cart for Availability


Select a Size

Change View
100 μG
$753.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$753.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

species reactivity

influenza A virus (avian)

concentration

1 mg/mL

technique(s)

ELISA: suitable

isotype

IgG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Avian Influenza Neuraminidase Antibody: Influenza A virus is a major public health threat, killing more than 30,000 people per year in the USA. Novel influenza virus strains emerge periodically to which humans have little or no immunity, resulting in devastating pandemics. Influenza A can exist in a variety of animals; however it is in birds that all subtypes can be found. These subtypes are classified based on the combination of the virus coat glycoproteins hemagglutinin (HA) and neuraminidase (NA) subtypes. During 1997, an H5N1 avian influenza virus was determined to be the cause of death in 6 of 18 infected patients in Hong Kong. There was some evidence of human to human spread of this virus, but it is thought that the transmission efficiency was fairly low. Although it has been known that cleavage site and glycosylation patterns of the HA protein play important roles in determining the pathogenicity of H5 avian influenza viruses, it has only recently been shown that an additional glycosylation site within the globular head of the NA protein also contributes to the high virulence of the H5N1 virus.

Immunogen

Avian influenza neuraminidase antibody was raised against a synthetic peptide corresponding to 15 amino acids near the carboxy terminus of the avian influenza neuraminidase protein.Efforts were made to use relatively conserved regions as the antigen.The immunogen is located within the last 50 amino acids of Avian Influenza Neuraminidase.

Application

Avian influenza neuraminidase antibody can be used for the detection of the avian influenza neuraminidase protein from the H5N1 strain of Avian influenza A in ELISA. It will detect 10 ng of free peptide at 1 μg/mL.

Physical form

Supplied in PBS containing 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service