Skip to Content
Merck
All Photos(3)

Key Documents

SAB3501043

Sigma-Aldrich

Anti-WFDC2 antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$532.00

$532.00


Check Cart for Availability


Select a Size

Change View
100 μG
$532.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$532.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

predicted mol wt 14 kDa

species reactivity

human

concentration

1 mg/mL

technique(s)

ELISA: suitable
immunoblotting: suitable
immunofluorescence: suitable
immunohistochemistry: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... WFDC2(10406)

Immunogen

Antibody was raised against a 16 amino acid peptide near the amino terminus of human WFDC2

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3501043.

Physical form

Supplied at 1 mg/mL in PBS with 0.02% sodim azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yao Chen et al.
Oncology reports, 29(1), 288-296 (2012-11-07)
The WAP four-disulfide core domain protein 2 (WFDC2) is frequently overexpressed in epithelial ovarian cancer cells and has been proposed as a potential biomarker. The biological function of WFDC2 in tumor progression remains unclear. In this study, the stable expression of short
Ronny Drapkin et al.
Cancer research, 65(6), 2162-2169 (2005-03-23)
Among the genes most commonly identified in gene expression profiles of epithelial ovarian carcinomas (EOC) is the gene for human epididymis protein 4 (HE4). To ascertain its clinical utility, we did a comprehensive assessment of HE4 protein expression in benign
Lynne Bingle et al.
Respiratory research, 7, 61-61 (2006-04-08)
The Whey Acidic Protein domain is an evolutionarily conserved motif found in a number of proteins, the best studied of which are antiproteinases involved in the innate immune defence of multiple epithelia. We recently characterised the WFDC2 gene which encodes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service