Skip to Content
Merck
All Photos(3)

Key Documents

SAB3500977

Sigma-Aldrich

Anti-ACE2 antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$626.00

$626.00


Available to ship on22 April 2025Details



Select a Size

Change View
100 μG
$626.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$626.00


Available to ship on22 April 2025Details


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

predicted mol wt 89 kDa

species reactivity

human

concentration

1 mg/mL

technique(s)

ELISA: suitable
immunoblotting: suitable
immunofluorescence: suitable
immunohistochemistry: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ACE2(59272)

General description

Angiotensin-converting enzyme-2 (ACE2), also referred as ACEH (human ACE homolog), is an integral membrane protein. It is encoded by the gene located at human chromosome Xp22.2. Ace 2 is found in variety of tissues, but the higher levels of protein is observed in vascular endothelium, lungs, heart, kidney, and testis. The protein contains a N-terminal single catalytic metallopeptidase unit, a C-terminal noncatalytic domain and a cytoplasmic tail.

Immunogen

Antibody was raised against a synthetic peptide corresponding to amino acids near the C-terminus of human ACE2.

Biochem/physiol Actions

Angiotensin-converting enzyme-2 (ACE2) as a carboxypeptidase cleaves angiotensin (Ang) I to Ang 1-9. In addition, it also degrades Ang II to Ang 1–7 and plays a vital role in regulation of blood pressure. Ace 2 aids in maintaining the balance of local renin-angiotensin system (RAS). It also helps in absorbing the neutral amino acids from the intestine and the kidney. It is a cellular receptor for spike glycoprotein of severe acute respiratory syndrome-coronavirus (SARS-CoV) and the new coronavirus (SARS-CoV-2), which is known to cause coronavirus disease 2019 (COVID-19). Downregulated expression of Ace 2 has been observed in patients with cardiovascular diseases.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500977.

Physical form

Supplied at 1 mg/mL in PBS with 0.02% sodim azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chris Tikellis et al.
International journal of peptides, 2012, 256294-256294 (2012-04-27)
Angiotensin-converting enzyme 2 (ACE2) shares some homology with angiotensin-converting enzyme (ACE) but is not inhibited by ACE inhibitors. The main role of ACE2 is the degradation of Ang II resulting in the formation of angiotensin 1-7 (Ang 1-7) which opposes
M Donoghue et al.
Circulation research, 87(5), E1-E9 (2000-09-02)
ACE2, the first known human homologue of angiotensin-converting enzyme (ACE), was identified from 5' sequencing of a human heart failure ventricle cDNA library. ACE2 has an apparent signal peptide, a single metalloprotease active site, and a transmembrane domain. The metalloprotease
ACE2 is on the X chromosome: could this explain COVID-19 gender differences?
Esther Culebras et al.
European heart journal (2020-06-25)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service