Skip to Content
Merck
All Photos(2)

Key Documents

SAB3500457

Sigma-Aldrich

Anti-TEM4 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-ARHGEF17, Anti-P164RHOGEF, Anti-Rho guanine nucleotide exchange factor 17, Anti-Tumor endothelial marker 4

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$541.00

$541.00


Check Cart for Availability


Select a Size

Change View
100 μG
$541.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$541.00


Check Cart for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

predicted mol wt 220 kDa

species reactivity

human

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ARHGEF17(9828)

General description

Rho guanine nucleotide exchange factor 17 (ARHGEF17), also known as tumor endothelial marker 4 (TEM4) and p164-RhoGEF (Rho-specific guanine-nucleotide exchange factor), encodes a guanine nucleotide exchange factor (GEF) for RhoA GTPases. The encoded protein consists of 1510 amino acids and contains a RhoGEF-specific Dbl homology (DH) domain. It does not contain the typical pleckstrin homology domain. TEM4 is predominantly expressed in the heart. Overexpression of this protein inhibits the transcription of M3 muscarinic receptor-induced genes. TEM4 modulates endothelial cell migration.

Immunogen

TEM4 antibody was raised against a 18 amino acid peptide near the center of human TEM4.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500457.

Physical form

Supplied in PBS with 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A mammalian Rho-specific guanine-nucleotide exchange factor (p164-RhoGEF) without a pleckstrin homology domain
Ulrich Rumenapp
BioChemistry: An Indian Journal (2002)
The RhoGEF TEM4 Regulates Endothelial Cell Migration by Suppressing Actomyosin Contractility.
Mitin N
PLoS ONE (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service