Skip to Content
Merck
All Photos(1)

Key Documents

SAB3500121

Sigma-Aldrich

Monoclonal Anti-ORAI3 antibody produced in mouse

clone 1B4F1, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-Calcium release-activated calcium channel protein 3, Anti-TMEM142C, Anti-Transmembrane protein 142C

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$556.00

$556.00


Estimated to ship on22 May 2025



Select a Size

Change View
100 μG
$556.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$556.00


Estimated to ship on22 May 2025


biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1B4F1, monoclonal

form

buffered aqueous solution

species reactivity

human, rat

technique(s)

indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ORAI3(93129)

Related Categories

Immunogen

raised against a 19 amino acid peptide from near the carboxy terminus of human ORAI3.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Antigen stimulation of immune cells triggers Ca++ entry t hrough Ca++ release-activated Ca++ (CRAC) channels. ORAI3 is one of two mammalian homologs to ORAI1, a recently identified four-transmembrane spanning protein that is an essential component of CRAC. All three homologs have been shown to function as Ca++ plasma membrane channels gated through interactions with STIM1, the store-activated endoplasmic reticulum Ca++ sensor. However, ORAI3 channels failed to produce detectable Ca++ selective currents in cells co-transfected with ORAI3 and STIM1, indicating that ORAI3 channels undergo a lesser degree of depotentiation than ORAI1 or ORAI2 (5). Na+ currents through ORAI1, 2 and 3 channels were equally inhibited by extracellular Ca++, indicating that each have similar affinities for Ca++ within the selectivity filter. This antibody is predicted to have no cross-reactivity to ORAI1 or ORAI2. Larger molecular weight bands are sometimes seen in SDS-PAGE; these may represent post-translationally modified ORAI 3.

Physical form

Supplied at approx. 1 mg/mL in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service