Skip to Content
Merck
All Photos(1)

Key Documents

SAB3500079

Sigma-Aldrich

Anti-PALB2 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-FANCN, Anti-Partner and localizer of BRCA2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$556.00

$556.00


Check Cart for Availability


Select a Size

Change View
100 μG
$556.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$556.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, rat, human

technique(s)

indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PALB2(79728)

General description

Partner and Localizer of BRCA2 (PALB2) gene with 13 exons is mapped to human chromosome 16p12.2. The gene codes for a 1,186 amino acid protein characterized with a coiled-coil motif at the N terminus and a C-terminal domain containing a series of WD repeats.

Immunogen

PALB2 antibody was raised against a 15 amino acid peptide from near the carboxy terminus of human PALB2.

Application

Anti-PALB2 antibody produced in rabbit has been used in western blot analysis.

Biochem/physiol Actions

Partner and Localizer of BRCA2 (PALB2) plays a vital role in BRCA2 (breast cancer 2, early onset)-related pathways. The breast cancer early onset proteins BRCA1 and BRCA2 are central to the repair of DNA damage by homologous recombination and are strongly associated with inherited breast and ovarian cancer. The protein PALB2 is required for the localization of BRCA2 to sites of DNA damage and like BRCA1 and 2 is a breast cancer susceptibility gene. PALB2 is thought to function through directly binding to BRCA1, which allows the PALB2 protein to organize BRCA2 and the recombinase RAD51 at the site of DNA damage. Analysis of the BRCA1-PALB2-BRCA2-RAD51 network may allow predictions to be made of the responsiveness of a tumor to therapeutic treatments. At least four isoforms of PALB2 are known to exist.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

The breast cancer early onset proteins BRCA1 and BRCA2 are central to the repair of DNA damage by homologous recombination and are strongly associated with inherited breast and ovarian cancer. The protein PALB2 is required for the localization of BRCA2 to sites of DNA damage and like BRCA1 and 2 is a breast cancer susceptibility gene. PALB2 is thought to function through directly binding to BRCA1, which allows the PALB2 protein to organize BRCA2 and the recombinase RAD51 at the site of DNA damage. Analysis of the BRCA1-PALB2-BRCA2-RAD51network may allow predictions to be made of the responsiveness of a particular tumor to therapeutic treatments. At least four isoforms of PALB2 are known to exist.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500079.

Physical form

Supplied at approx. 1 mg/mL in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A cell-penetrating antibody inhibits human RAD51 via direct binding
Turchick A, et al.
Nucleic Acids Research, 45, 11782-11799 (2017)
PALB2/FANCN: Recombining Cancer and Fanconi Anemia
Tischkowitz M, et al.
Cancer research, 70, 7353-7359 (2010)
Audrey Turchick et al.
Nucleic acids research, 45(20), 11782-11799 (2017-10-17)
RAD51, a key factor in homology-directed repair (HDR), has long been considered an attractive target for cancer therapy, but few specific inhibitors have been found. A cell-penetrating, anti-DNA, lupus autoantibody, 3E10, was previously shown to inhibit HDR, sensitize tumors to
PALB2 mutations in German and Russian patients with bilateral breast cancer
Bogdanova N, et al.
Breast Cancer Research and Treatment, 126, 545-550 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service