Skip to Content
Merck
All Photos(2)

Key Documents

SAB3500065

Sigma-Aldrich

Anti-Seasonal H1N1 Neuraminidase antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Flu N1, Anti-Influenza N1, Anti-NA, Anti-Seasonal Influenza A (H1N1) Neuraminidase

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$556.00

$556.00


Check Cart for Availability


Select a Size

Change View
100 μG
$556.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$556.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

viral

technique(s)

indirect ELISA: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Immunogen

Neuraminidase antibody was raised against a synthetic peptide from the seasonal influenza (H1N1) Neuraminidase protein. This antibody is a cognate pair with product SAB3500066.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Influenza A virus is a major public health threat, killing more than 30,000 people per year in the USA (1). In early 2009, a novel swine-origin influenza A (H1N1) virus was identified in specimens obtained from patients in Mexico and the United States (2). The virus spread quickly around the world and on June 11, 2009, the World Health Organization declared it a pandemic (3). Influenza A virus has one of sixteen possible Hemagglutinin (HA) surface proteins and one of nine possible neuraminidase (NA) surface proteins. The Hemagglutinin protein facilitates viral attachment while neuraminidase is involved in viral release. These proteins also elicit immune responses that prevent infection or independently reduce viral replication. The genetic make-up of this swine flu virus is unlike any other: it is an H1N1 strain that combines a triple assortment first identified in 1998 including human, swine, and avian influenza with two new pig H3N2 virus genes from Eurasia, themselves of recent human origin (4). The distinct antigenic properties of the new swine influenza virus compared with seasonal influenza A (H1N1) virus suggest that human immunity against new swine influenza virus is limited, although the age distribution of reported cases suggests some degree of protection in older age groups (5). This antibody is specific for Neuraminidase from the seasonal influenza (H1N1) and will not recognize the corresponding Neuraminidase sequence from the swine-origin influenza virus (A/California/14/2009 (H1N1)).

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500065.

Physical form

Supplied at approx. 1 mg/mL in phosphate buffered saline containing 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service