Skip to Content
Merck
All Photos(2)

Key Documents

SAB3500051

Sigma-Aldrich

Anti-SHOC2 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Leucine-rich repeat protein SHOC-2, Anti-Ras-binding protein Sur-8, Anti-SOC2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$491.00

$491.00


Estimated to ship on29 May 2025



Select a Size

Change View
100 μG
$491.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$491.00


Estimated to ship on29 May 2025


biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat, mouse, human

technique(s)

immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SHOC2(8036)

General description

Leucine-rich repeat protein SHOC-2 is a widely expressed protein composed almost entirely of leucine-rich repeats (LRR), with a lysine-rich sequence at the amino terminus and is cytoplasmically localized. SHOC2 acts as a positive modulator of the RAS-MAPK signaling cascade.

Immunogen

SHOC2 antibody was raised against a 21 amino acid peptide near the amino terminus of human SHOC2.

Biochem/physiol Actions

Leucine-rich repeat protein SHOC2 participates in protein binding / transferase activity in the fibroblast growth factor receptor signaling pathway and Ras protein signal transduction. SHOC2, together with protein phosphatase 1c (PP1c), forms a highly specific M-Ras effector complex and is essential for the activation of MAPK pathway by growth factors. Furthermore, in tumor cells with Ras gene mutations, inhibition of SHOC2 expression inhibits MAPK, but not PI3K activity. The SHOC2-PP1c holoenzyme provides an attractive therapeutic target for inhibition of the MAPK pathway in cancer. Recent studies show that aberrantly acquired N-myristoylation of SHOC2 causes human disease Noonan-like syndrome with loose anagen hair.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500051.

Physical form

Supplied at approx. 1 mg/mL in phosphate buffered saline containing 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mutation in SHOC2 promotes aberrant protein N-myristoylation and underlies Noonan-like syndrome with loose anagen hair
Viviana Cordeddu
Nature Genetics, 41(9), 1022-1026 (2009)
Noonan syndrome-like disorder with loose anagen hair: a second case with neuroblastoma.
Garavelli L
American Journal of Medical Genetics. Part A, 167A(8), 1902-1907 (2015)
soc-2 encodes a leucine-rich repeat protein implicated in fibroblast growth factor receptor signaling
Laura M
Proceedings of the National Academy of Sciences of the USA, 95(12), 6903-6908 (1998)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service