Skip to Content
Merck
All Photos(2)

Key Documents

SAB2900288

Sigma-Aldrich

Anti-TICAM1/TRIF (AB1) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$645.00

$645.00


Check Cart for Availability


Select a Size

Change View
50 μG
$645.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$645.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TICAM1(148022)

General description

Toll like receptor adaptor molecule 1 (TICAM1) is ubiquitously expressed and possesses an N-terminal proline-rich domain, a TIR (Toll/interleukin-1 receptor) motif and a C-terminal proline-rich domain. The gene encoding this protein is localized on human chromosome 19p13.3.

Immunogen

Synthetic peptide from human TICAM1 / TRIF.

Biochem/physiol Actions

Toll like receptor adaptor molecule 1 (TICAM1) plays a crucial role as an adaptor molecule and links toll-like receptor 3 (TLR3) to interferon-β (IFN-β) promoter activation. It is also involved in IFN-β production. TICAM-1 pathway has an essential role in host protection and innate immune responses against poliovirus (PV) infection. Mutations in the gene have been associated with the development of papillary thyroid cancer, follicular thyroid cancer and differentiated thyroid cancer.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The TLR3/TICAM-1 pathway is mandatory for innate immune responses to poliovirus infection.
Oshiumi H
Journal of Immunology (2011)
Selected single-nucleotide polymorphisms in FOXE1, SERPINA5, FTO, EVPL, TICAM1 and SCARB1 are associated with papillary and follicular thyroid cancer risk: replication study in a German population.
Sigurdson AJ
Carcinogenesis (2016)
TICAM-1, an adaptor molecule that participates in Toll-like receptor 3-mediated interferon-beta induction.
Oshiumi H
Nature Immunology (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service