Skip to Content
Merck
All Photos(4)

Documents

SAB2702236

Sigma-Aldrich

Monoclonal Anti-NuMA antibody produced in mouse

clone GT3611, affinity isolated antibody

Synonym(s):

NMP-22, NUMA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

GT3611, monoclonal

form

buffered aqueous solution

mol wt

238kDa

species reactivity

human

concentration

1mg/mL

technique(s)

indirect immunofluorescence: suitable
western blot: 500-3000

isotype

IgG2b

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NUMA1(4926)

Immunogen

Recombinant protein encompassing a sequence within the N-terminus region of human NuMA.

Application

Suggested starting dilutions are as follows: ICC/IF: 1:100-1:1000, WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Other Notes

Purification: Affinity purified by Protein G

Physical form

Phosphate-buffered saline, no preservative added.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service