Skip to Content
Merck
All Photos(2)

Key Documents

SAB2702133

Sigma-Aldrich

Monoclonal Anti-CEP164 antibody produced in mouse

Synonym(s):

Cep164 Antibody, Cep164 Antibody - Monoclonal Anti-CEP164 antibody produced in mouse, NPHP15

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$513.00

$513.00


Check Cart for Availability


Select a Size

Change View
100 μL
$513.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$513.00


Check Cart for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

13
monoclonal

form

liquid

species reactivity

human

concentration

1 mg/mL

technique(s)

immunocytochemistry: suitable
immunofluorescence: suitable
immunoprecipitation (IP): suitable
western blot: 1:500-1:3000

isotype

IgG1

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CEP164(22897)

General description

The centrosomal protein 164 (CEP164) is expressed at the distal ends of the mother centriole. This protein consists of an N-terminal WW domain and three coiled-coil regions. The CEP164 gene is located on the human chromosome at 11q23.3.

Immunogen

Human Cep164

Application

Monoclonal Anti-CEP164 antibody produced in mouse has been used in immunoblotting (5 μg/ml).[1]

Biochem/physiol Actions

The centrosomal protein 164 (CEP164) plays an important role in the synthesis of the primary cilium. This protein interacts with several proteins during ciliogenesis such as Rabin-8, Tau tubulin kinase‐2 (TTBK2), coiled-coil domain containing 41 (CCDC41), and dishevelled (DVL). CEP164 protein is also involved in G2/M checkpoint and nuclear divisions. This protein is a key player in the DNA damage-activated ataxia telangiectasia mutated (ATM)/Rad3-related (ATR) signaling cascade since it is required for the proper phosphorylation of the histone family member X (H2AX), replication protein A (RPA), checkpoint kinase 1 and 2 (CHK1/2). Mutations in the CEP164 gene leads to a condition known as nephronophthisis (NPHP)‐related ciliopathies.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Phosphate-buffered saline, no preservative added.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Toshiaki Oda et al.
Genes to cells : devoted to molecular & cellular mechanisms, 19(12), 927-940 (2014-10-10)
Primary cilia are formed by extending the microtubule-based axoneme from the mother centriole-derived basal body. Recruitment of Tau tubulin kinase-2 (TTBK2) to the mother centriole and subsequent removal of CP110 and its interactor Cep97 are crucial for the initiation of
Susanne Graser et al.
The Journal of cell biology, 179(2), 321-330 (2007-10-24)
Primary cilia (PC) function as microtubule-based sensory antennae projecting from the surface of many eukaryotic cells. They play important roles in mechano- and chemosensory perception and their dysfunction is implicated in developmental disorders and severe diseases. The basal body that
Sudhakar Sivasubramaniam et al.
Genes & development, 22(5), 587-600 (2008-02-20)
The activation of the ataxia telangiectasia mutated (ATM) and ATM/Rad3-related (ATR) kinases triggers a diverse cellular response including the initiation of DNA damage-induced cell cycle checkpoints. Mediator of DNA Damage Checkpoint protein, MDC1, and H2AX are chromatin remodeling factors required
Rebecca V Walker et al.
Nature communications, 10(1), 4072-4072 (2019-09-08)
The human PKD2 locus encodes Polycystin-2 (PC2), a TRPP channel that localises to several distinct cellular compartments, including the cilium. PKD2 mutations cause Autosomal Dominant Polycystic Kidney Disease (ADPKD) and affect many cellular pathways. Data underlining the importance of ciliary

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service