Skip to Content
Merck
All Photos(11)

Key Documents

SAB2701077

Sigma-Aldrich

Anti-CS antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

CS, citrate synthase

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$604.00

$604.00


Available to ship on14 April 2025Details



Select a Size

Change View
100 μL
$604.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$604.00


Available to ship on14 April 2025Details


biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

chicken, monkey, human, mouse, rat

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable
indirect immunofluorescence: suitable
western blot: 500-3000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CS(1431)

Immunogen

Recombinant fragment corresponding to a region within amino acids 110 and 412 of Citrate synthetase according to NP_938083

Application

Suggested starting dilutions are as follows: ICC/IF: 1:100-1:1000, IHC-P: 1:100-1:1000, IP: 1:100-1:500, WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Biochem/physiol Actions

The protein encoded by this gene is a Krebs tricarboxylic acid cycle enzyme that catalyzes the synthesis of citrate from oxaloacetate and acetyl coenzyme A. The enzyme is found in nearly all cells capable of oxidative metablism. This protein is nuclear encoded and transported into the mitochondrial matrix, where the mature form is found. [provided by RefSeq]

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

1XPBS, 20% Glycerol (pH7). 0.025% ProClin 300 was added as a preservative.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joseph J Bass et al.
The Journal of physiology, 599(3), 963-979 (2020-12-02)
Reduced vitamin D receptor (VDR) expression prompts skeletal muscle atrophy. Atrophy occurs through catabolic processes, namely the induction of autophagy, while anabolism remains unchanged. In response to VDR-knockdown mitochondrial function and related gene-set expression is impaired. In vitro VDR knockdown
Siva R Uppalapati et al.
Vaccine, 32(25), 3075-3081 (2014-04-05)
Clostridium perfringens and Staphylococcus aureus are the two important bacteria frequently associated with majority of the soft tissue infections. The severity and progression of the diseases caused by these pathogens are attributed primarily to the alpha toxins they produce. Previously
Stephen P Ashcroft et al.
American journal of physiology. Cell physiology, 318(3), C536-C541 (2020-01-16)
Vitamin D deficiency has been linked to a reduction in skeletal muscle function and oxidative capacity; however, the mechanistic bases of these impairments are poorly understood. The biological actions of vitamin D are carried out via the binding of 1α,25-dihydroxyvitamin
Katie A O'Brien et al.
Biochimica et biophysica acta. Molecular basis of disease, 1865(4), 844-853 (2018-07-29)
Hypoxia is a feature of many disease states where convective oxygen delivery is impaired, and is known to suppress oxidative metabolism. Acclimation to hypoxia thus requires metabolic remodelling, however hypoxia tolerance may be aided by dietary nitrate supplementation. Nitrate improves
Ben Stocks et al.
Frontiers in physiology, 8, 941-941 (2017-12-20)
Tumour protein 53 (p53) has been implicated in the regulation of mitochondrial biogenesis in skeletal muscle, with whole-body p53 knockout mice displaying impairments in basal mitochondrial content, respiratory capacity, and enzyme activity. This study aimed to determine the effect of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service