Skip to Content
Merck
All Photos(5)

Key Documents

SAB2700882

Sigma-Aldrich

Anti-AKR1C1 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

2-ALPHA-HSD, 20-ALPHA-HSD, AKR1C1, C9

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$513.00

$513.00


Estimated to ship on30 April 2025



Select a Size

Change View
100 μL
$513.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$513.00


Estimated to ship on30 April 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, human

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
western blot: 500-3000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... AKR1C1(1645)

Immunogen

Recombinant fragment corresponding to a region within amino acids 22 and 248 of AKR1C1 according to NP_001344

Application

Suggested starting dilutions are as follows: ICC/IF: 1:100-1:1000, IHC: Assay-dependent dilution, IHC-P: 1:100-1:1000, WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Biochem/physiol Actions

This gene encodes a member of the aldo/keto reductase superfamily, which consists of more than 40 known enzymes and proteins. These enzymes catalyze the conversion of aldehydes and ketones to their corresponding alcohols by utilizing NADH and/or NADPH as cofactors. The enzymes display overlapping but distinct substrate specificity. This enzyme catalyzes the reaction of progesterone to the inactive form 20-alpha-hydroxy-progesterone. This gene shares high sequence identity with three other gene members and is clustered with those three genes at chromosome 10p15-p14. [provided by RefSeq]

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

0.1M Tris, 0.1M Glycine, 10% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xianjun Wang et al.
Molecular medicine reports, 24(6) (2021-10-01)
Acute lung injury (ALI) is a respiratory tract disease characterized by increased alveolar/capillary permeability, lung inflammation and structural damage to lung tissues, which can progress and transform into acute respiratory distress syndrome (ARDS). Although there are several treatment strategies available
Naoki Mine et al.
Molecular cancer therapeutics, 13(9), 2215-2225 (2014-07-24)
CBP501 is an anticancer drug candidate that was investigated in two randomized phase II clinical trials for patients with nonsquamous non-small cell lung cancer (NSCLC) and malignant pleural mesothelioma (MPM). CBP501 has been shown to have two mechanisms of action

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service