Skip to Content
Merck
All Photos(4)

Key Documents

SAB2700755

Sigma-Aldrich

Anti-BMP4 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

BMP2B, BMP2B1, BMP4, MCOPS6

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$604.00

$604.00


Check Cart for Availability


Select a Size

Change View
100 μL
$604.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$604.00


Check Cart for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, human

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
western blot: 500-3000

NCBI accession no.

UniProt accession no.

application(s)

research pathology

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... BMP4(652)

Immunogen

Recombinant fragment corresponding to a region within amino acids 1 and 287 of BMP4 according to NP_001193

Application

Suggested starting dilutions are as follows: ICC/IF: 1:100-1:1000, IHC-P: 1:100-1:1000, WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Biochem/physiol Actions

The protein encoded by this gene is a member of the bone morphogenetic protein family which is part of the transforming growth factor-beta superfamily. The superfamily includes large families of growth and differentiation factors. Bone morphogenetic proteins were originally identified by an ability of demineralized bone extract to induce endochondral osteogenesis in vivo in an extraskeletal site. This particular family member plays an important role in the onset of endochondral bone formation in humans, and a reduction in expression has been associated with a variety of bone diseases, including the heritable disorder Fibrodysplasia Ossificans Progressiva. Alternative splicing in the 5′ untranslated region of this gene has been described and three variants are described, all encoding an identical protein. [provided by RefSeq]

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

0.1M Tris, 0.1M Glycine, 10% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaopeng Hu et al.
Scientific reports, 5, 12982-12982 (2015-08-13)
The male's ability to reproduce is completely dependent on Sertoli cells. However, the mechanisms governing the functional integrity of Sertoli cells have remained largely unexplored. Here, we demonstrate that deletion of Shp2 in Sertoli cells results in infertility in mice.
Alex Yuri Simões Sato et al.
Atherosclerosis, 235(1), 45-55 (2014-05-13)
Monocytes and macrophages, together with vascular smooth muscle cells (VSMCs), play key roles at all stages of atherogenesis. There is also growing evidence that BMP signaling is involved in vascular diseases, including atherosclerosis. Here we evaluate the role played by
Tamás Juhász et al.
Journal of molecular neuroscience : MN, 54(3), 555-573 (2014-08-13)
Presence of the pituitary adenylate cyclase-activating polypeptide (PACAP) signalling has been proved in various peripheral tissues. PACAP can activate protein kinase A (PKA) signalling via binding to pituitary adenylate cyclase-activating polypeptide type I receptor (PAC1), vasoactive intestinal polypeptide receptor (VPAC)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service