SAB2700083
Anti-CKMT1A antibody produced in rabbit
affinity isolated antibody, buffered aqueous solution
Synonym(s):
CKMT1, CKMT1A, CKMT1B, FLJ50967
About This Item
Recommended Products
biological source
rabbit
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
species reactivity
human
technique(s)
immunohistochemistry: suitable
western blot: 500-3000
NCBI accession no.
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... CKMT1A(548596)
Immunogen
Application
Biochem/physiol Actions
Features and Benefits
Physical form
Disclaimer
![](/deepweb/assets/sigmaaldrich/product/images/242/317/89e134be-e488-4e6d-9500-3baa2929e5c3/300/89e134be-e488-4e6d-9500-3baa2929e5c3.jpg)
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service