Skip to Content
Merck
All Photos(1)

Key Documents

SAB2501668

Sigma-Aldrich

Anti-UCHL1 (C-terminal) antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

PARK5, PGP9.5, PGP95, ubiquitin carboxyl-terminal esterase L1 (ubiquitin thiolesterase)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$447.00

$447.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1664


Select a Size

Change View
100 μG
$447.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$447.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1664

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

canine, bovine, rat, rabbit, pig, human

technique(s)

immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... UCHL1(7345)

General description

Ubiquitin C-terminal hydrolase L1 (UCHL1) is a deubiquitinase which is part of the ubiquitin proteasome system (UPS). The protein is expressed in the nervous system and gonads. The gene encoding UCHL1 is localized on human chromosome 4p13.

Immunogen

Peptide with sequence EVRFSAVALCK, from the C-terminal region of the protein sequence according toNP_004172.2

Biochem/physiol Actions

Ubiquitin C-terminal hydrolase L1 (UCHL1) has a role in epithelial-mesenchymal transition. It also has roles in invasion and in the cell cycle. UCHL1 catalyzes the hydrolysis of carboxy-terminal ubiquitin esters and amides. The protein has been associated with various cancers.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in 20mM Tris (pH 7.3) and 150mM NaCl with 0.02% sodium azide and 0.5% bovine serum albumin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

UCH-LI acts as a novel prognostic biomarker in gastric cardiac adenocarcinoma.
YAng H
International Journal of Clinical and Experimental Pathology (2015)
Ubiquitin carboxyl-terminal esterase L1 (UCHL1) is associated with stem-like cancer cell functions in pediatric high-grade glioma.
Sanchez-diaz PC
PLoS ONE (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service