Skip to Content
Merck
All Photos(1)

Key Documents

SAB2501420

Sigma-Aldrich

Anti-PEBP4 (C-terminal) antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-CORK-1, Anti-CORK1, Anti-GWTM1933, Anti-MGC22776, Phosphatidylethanolamine-binding protein 4

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$417.00

$417.00


Check Cart for Availability


Select a Size

Change View
100 μG
$417.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$417.00


Check Cart for Availability

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PEBP4(157310)

General description

The gene PEBP4 (phosphatidylethanolamine binding protein 4) is mapped to human chromosome 8p21.3. It belongs to the PEBP family of proteins. The protein has potential glycosylation sites and can be secreted. It is mainly expressed in the skeletal muscle, heart and thyroid.

Immunogen

Peptide with sequence RERASEPKHKNQAE, from the C-terminal region of the protein sequence according to NP_659399.2

Biochem/physiol Actions

PEBP4 (phosphatidylethanolamine binding protein 4) regulates apoptosis by associating with Raf1 (proto-oncogene serine/threonine-protein kinase) and MEK1 (MAPK/ERK kinase 1). This association blocks TNFα (tumor necrosis α) or TRAIL (tumor necrosis factor-related apoptosis-inducing ligand) - mediated activation of MEK/ERK (extracellular signal-regulated kinase). PEBP4 is also involved in myoblast differentiation. Additionally, it promotes Akt (RAC-α serine/threonine-protein kinase) activation. The gene is expressed in various cancers and is associated with cancer progression. The PEBP4 mRNA is a target of microRNA miR-34a.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in 20mM Tris (pH 7.3) and 150mM NaCl with 0.02% sodium azide and 0.5% bovine serum albumin.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li Chushi et al.
Gene, 587(1), 42-47 (2016-04-18)
Alteration of metabolic profile is one of the hallmarks of cancer cells. Statin, the inhibitors for synthesis of cholesterol, has shown anti-cancer effects on the gastric cancer cells. However, the functions of its target, HMGCR, in the progression of gastric
Huan He et al.
Biochimica et biophysica acta, 1863(7 Pt A), 1682-1689 (2016-04-02)
Phosphatidylethanolamine binding proteins (PEBP) represent a superfamily of proteins that are conserved from bacteria to humans. In mammals, four members have been identified, PEBP1-4. To determine the functional differences among PEBP1-4 and the underlying mechanism for their actions, we performed
Guiping Yu et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 35(10), 10341-10349 (2014-07-21)
The aim of this study was to determine the relationship and underlying mechanisms between ectopic expression of phosphatidylethanolamine-binding protein 4 (PEBP4) and cisplatin (DDP)-induced cytotoxicity in the lung cancer cell line A549 to provide an experimental basis for future chemotherapeutic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service