Skip to Content
Merck
All Photos(1)

Key Documents

SAB2501226

Sigma-Aldrich

Anti-UNC5C antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-UNC5H3, unc-5 homolog C (C. elegans)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$383.00

$383.00


Check Cart for Availability


Select a Size

Change View
100 μG
$383.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$383.00


Check Cart for Availability

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, canine, human

technique(s)

immunohistochemistry: suitable
indirect ELISA: suitable
indirect immunofluorescence: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... UNC5C(8633)

Immunogen

Peptide with sequence NCTVSEEPTGID, from the internal region of the protein sequence according to NP_003719.2.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service