Skip to Content
Merck
All Photos(1)

Key Documents

SAB2500745

Sigma-Aldrich

Anti-PAM/PAL antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-PAL, Anti-PHM, Anti-Pancreatic peptidylglycine α-amidating monooxygenase, Anti-Peptidyl α-amidating enzyme, Anti-Peptidylglycine α-amidating monooxygenase

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$412.00

$412.00


Estimated to ship on24 May 2025



Select a Size

Change View
100 μG
$412.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

$412.00


Estimated to ship on24 May 2025


biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PAM(5066)

General description

Peptidylglycine α-amidating monooxygenase (PAM) is expressed in endocrine cells and neurons. It consists of two enzyme activities- peptidyl-glycine α-hydroxylating monoxygenase (PHM) and peptidyl-α-hydroxyglycine amidating lyase (PAL). The PAM gene is localized on human chromosome 5q21.1.

Immunogen

Peptide with sequence C-DQEKEDDGSESEE from the C Terminus of the protein sequence according to NP_000910.2; NP_620121.1; NP_620176.1; NP_620177.1.

Biochem/physiol Actions

Peptidylglycine α-amidating monooxygenase (PAM) has a role in the biosynthesis of bioactive peptides. It is involved in C-terminal α-amidation, which influences the activity of regulatory peptides of the respiratory tract.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The endocytic pathways of a secretory granule membrane protein in HEK293 cells: PAM and EGF traverse a dynamic multivesicular body network together.
Back N, et al.
European Journal of Cell Biology, 96(5), 407-417 (2017)
Peptidylglycine alpha-amidating monooxygenase: a multifunctional protein with catalytic, processing, and routing domains.
Eipper BA, et al.
Protein Science, 2(4), 489-497 (1993)
Distribution of peptidyl-glycine alpha-amidating mono-oxygenase (PAM) enzymes in normal human lung and in lung epithelial tumors.
Saldise L, et al.
The Journal of Histochemistry and Cytochemistry, 44(1), 3-12 (1996)
Localization of the gene encoding peptidylglycine alpha-amidating monooxygenase (PAM) to human chromosome 5q14-5q21.
Ouafik LH, et al.
Genomics, 18(2), 319-321 (1993)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service