Skip to Content
Merck
All Photos(1)

Key Documents

SAB2500713

Sigma-Aldrich

Anti-OCT4/POU5F1 antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-HGNC:9221, Anti-OTF4

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$467.00

$467.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1101


Select a Size

Change View
100 μG
$467.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

$467.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1101

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, mouse, canine, rat

technique(s)

indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... POU5F1(5460)

Immunogen

Peptide with sequence C-VTTLGSPMHSN from the C Terminus of the protein sequence according to NP_002692.2; NP_976034.2.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qing-Zhen Liu et al.
Experimental and therapeutic medicine, 12(3), 1651-1656 (2016-09-03)
Adult stem cells have been well characterized in numerous organs, with the exception of the kidneys. Therefore, the present study aimed to identify and isolate kidney-derived stem cells. A total of 12 Fischer 344 transgenic rats expressing the human diphtheria
Ying Jing et al.
Experimental and therapeutic medicine, 13(1), 280-284 (2017-01-27)
The molecular mechanisms underlying translationally-controlled tumor protein (TCTP) in the activation of octamer-binding transcription factor 4 (Oct-4) in kidney-derived stem cells have not been characterized. The aim of the present study was to identify the transcriptional activation of Oct-4 by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service