The epitope for this antibody has not been mapped. The immunogen used is a peptide with the sequence C-TQERVNNSNKKE, derived from the internal region of the protein sequence according to Accession Numbers NP_000823.4; NP_067544.1; NP_015566.1.
SAB2500698
Anti-NMDA Receptor 1/GRIN1 antibody produced in goat
affinity isolated antibody, buffered aqueous solution
Synonym(s):
Anti-N-methyl D-aspartate 1, Anti-NMDA1
Select a Size
$413.00
Select a Size
About This Item
$413.00
Recommended Products
biological source
goat
Quality Level
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
species reactivity
human, mouse
technique(s)
ELISA: suitable
immunofluorescence: suitable
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... GRIN1(2902)
Immunogen
Features and Benefits
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
Can epitope information, such as the location, be provided for SAB2500698?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service