Skip to Content
Merck
All Photos(1)

Key Documents

SAB2500409

Sigma-Aldrich

Anti-FOXA2/HNF3B antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Forkhead box A2, Anti-HNF3B, Anti-Hepatic nuclear factor-3-β, Anti-MGC19807, Anti-TCF3B

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$467.00

$467.00


Check Cart for Availability


Select a Size

Change View
100 μG
$467.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$467.00


Check Cart for Availability

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, mouse, rat

technique(s)

immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... FOXA2(3170)

General description

Forkhead box A2 (FOXA2), also known as hepatocyte nuclear factor 3-β (HNF3-β), is encoded by the gene mapped to human chromosome 20p11.21. It is expressed in the liver, pancreas, intestine, and lungs. FOXA2 is a member of forkhead box (FOX) gene family and is characterized by a DNA binding “winged helix domain”.[1]

Immunogen

Peptide with sequence C-GVYSRPIMNSS from the C Terminus of the protein sequence according to NP_068556.1; NP_710141.1.

Biochem/physiol Actions

Forkhead box A2 (FOXA2) or HNF3β is associated with embryonic formation of the primitive streak and endoderm. It also plays an important role in airway epithelial differentiation and is extensively expressed in type II pneumocytes. Decreased expression of FOXA2 is observed in lung cancer and non-small-cell lung cancer (NSCLC). FOXA2 plays a vital role in normal pancreatic β-cell function and aberration in the expression of the gene leads to hyperinsulinemic hypoglycemia. It also has an essential role in pancreatic α-cell differentiation. FOXA2 acts a regulator of the network of genes associated with intestinal epithelial cell function. Increased expression of FOXA2 in MKN-45 cells (human gastric cancer cell line) elevates E-cadherin expression and inhibits gastric cancer cell migration and invasion and therefore, FOXA2 is expressed at low levels in gastric adenocarcinoma tissues.[1][2][3][4]

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Down-expression of FOXA2 in gastric adenocarcinoma].
Zhang Z
Xi bao yu fen zi mian yi xue za zhi = Chinese journal of cellular and molecular immunology (2015)
Foxa2 is required for the differentiation of pancreatic alpha-cells.
Developmental Biology (2005)
FOXA2 regulates a network of genes involved in critical functions of human intestinal epithelial cells.
Gosalia N
Physiological Genomics (2015)
Frequent downregulation of the transcription factor Foxa2 in lung cancer through epigenetic silencing.
Basseres DS
Lung Cancer (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service