Skip to Content
Merck
All Photos(1)

Key Documents

SAB2500302

Sigma-Aldrich

Anti-DDAH2 antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-DDAHII, Anti-Dimethylarginine dimethylaminohydrolase 2, Anti-G6a, Anti-NG30

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$507.00

$507.00


Check Cart for Availability


Select a Size

Change View
100 μG
$507.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$507.00


Check Cart for Availability

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, human, rat

technique(s)

indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DDAH2(23564)

General description

DDAH (Dimethylarginine dimethylaminohydrolase) exists as two isoforms; DDAH1, and DDAH2. Tissues expressing endothelial nitric oxide synthases is predominated by DDAH2.

Immunogen

Peptide with sequence SSLCLVLSTRPHS from the C Terminus of the protein sequence according to NP_039268.1.

Application

Anti-DDAH2 antibody produced in goat is suitable for indirect ELISA and western blot applications.

Biochem/physiol Actions

DDAH2 (Dimethylarginine dimethylaminohydrolase 2) Activity partly determines the cellular concentration of methylarginine. Concentration of ADMA (asymmetrical dimethylarginine) in endothelial cells is increased on the advent of pharmacological inhibition of DDAH, which also leads to the inhibition of NO-mediated endothelium dependent relaxation of blood vessels.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J M Leiper et al.
The Biochemical journal, 343 Pt 1, 209-214 (1999-09-24)
Methylarginines inhibit nitric oxide synthases (NOS). Cellular concentrations of methylarginines are determined in part by the activity of dimethylarginine dimethylaminohydrolase (DDAH; EC 3.5.3. 18). We have cloned human DDAH and identified and expressed a second novel DDAH isoform (DDAH I

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service