Skip to Content
Merck
All Photos(1)

Key Documents

SAB2109093

Sigma-Aldrich

Anti-Sgms1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

9530058O11Rik, AI841905, C80702, MGC30540, Mob

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$563.00

$563.00


Check Cart for Availability


Select a Size

Change View
100 μL
$563.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$563.00


Check Cart for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

46 kDa

species reactivity (predicted by homology)

canine, mouse, bovine, rabbit, human, guinea pig, rat, horse

concentration

0.5 mg/mL

technique(s)

western blot: 1 μg/mL

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SGMS1(259230)

General description

Bidirectional lipid cholinephosphotransferase is capable of converting phosphatidylcholine (PC) and ceramide to sphingomyelin (SM) and diacylglycerol (DAG) and vice versa. Direction is dependent on the relative concentrations of DAG and ceramide as phosphocholine acceptors. Sgms1 directly and specifically recognizes the choline head group on the substrate. It also requires two fatty chains on the choline-P donor molecule in order to be recognized efficiently as a substrate. Sgms1 does not function strictly as a SM synthase and suppresses BAX-mediated apoptosis and also prevents cell death in response to stimuli such as hydrogen peroxide, osmotic stress, elevated temperature and exogenously supplied sphingolipids. Sgms1 may protect against cell death by reversing the stress-inducible increase in levels of proapoptotic ceramide.

Immunogen

Synthetic peptide directed towards the middle region of Mouse Sgms1

Sequence

Synthetic peptide located within the following region: LTTVMISVVHERVPPKEVQPPLPDTFFDHFNRVQWAFSICEINGMILVGL

Physical form

Supplied at 0.5 mg/ml in phosphate-buffered saline, 0.09% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service