Skip to Content
Merck
All Photos(3)

Key Documents

SAB2108745

Sigma-Aldrich

Anti-KLF6

affinity isolated antibody

Synonym(s):

Anti- COPEB, Anti- CPBP, Anti- GBF, Anti-BCD1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$677.00

$677.00


Check Cart for Availability


Select a Size

Change View
100 μL
$677.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$677.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

32 kDa

species reactivity

guinea pig, mouse, bovine, horse, rat, dog, human, rabbit

concentration

0.5-1 mg/mL

technique(s)

immunoblotting: suitable

accession no.

NM_001300

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KLF6(1316)

Immunogen

Synthetic peptide directed towards the N terminal region of human KLF6

Biochem/physiol Actions

KLF6 is a nuclear protein that has three zinc fingers at the end of its C-terminal domain, a serine/threonine-rich central region, and an acidic domain lying within the N-terminal region. The zinc fingers of this protein are responsible for the specific DNA binding with the guanine-rich core promoter elements. The central region might be involved in activation or posttranslational regulatory pathways, and the acidic N-terminal domain might play an important role in the process of transcriptional activation. It is capable of activating transcription approximately 4-fold either on homologous or heterologous promoters. KLF6 may participate in the regulation and/or maintenance of the basal expression of pregnancy-specific glycoprotein genes and possibly other TATA box-less genes.This gene encodes a nuclear protein that has three zinc fingers at the end of its C-terminal domain, a serine/threonine-rich central region, and an acidic domain lying within the N-terminal region. The zinc fingers of this protein are responsible for the specific DNA binding with the guanine-rich core promoter elements. The central region might be involved in activation or posttranslational regulatory pathways, and the acidic N-terminal domain might play an important role in the process of transcriptional activation. It is capable of activating transcription approximately 4-fold either on homologous or heterologous promoters. The DNA binding and transcriptional activity of this protein, in conjunction with its expression pattern, suggests that this protein may participate in the regulation and/or maintenance of the basal expression of pregnancy-specific glycoprotein genes and possibly other TATA box-less genes. Two transcript variants encoding the same protein have been found for this gene. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications. PRIMARYREFSEQ_SPAN PRIMARY_IDENTIFIER PRIMARY_SPAN COMP 1-731 BM544849.1 20-750 732-1504 BC000311.2 669-1441 1505-1598 BC004301.1 1440-1533

Sequence

Synthetic peptide located within the following region: REKKEESELKISSSPPEDTLISPSFCYNLETNSLNSDVSSESSDSSEELS

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jing Han et al.
Gerontology, 68(10), 1145-1165 (2022-05-17)
Sepsis is a life-threatening inflammatory state that can result in septic acute kidney injury (SAKI). Circular RNAs (circRNAs) are implicated in various inflammatory diseases including SAKI. This study investigated the effect of circHIPK3 on inflammatory responses and cell senescence in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service