Skip to Content
Merck
All Photos(2)

Key Documents

SAB2108487

Sigma-Aldrich

Anti-BTG2

affinity isolated antibody

Synonym(s):

Anti- MGC126064, Anti- PC3, Anti- TIS21, Anti-MGC126063

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$797.00

$797.00


Check Cart for Availability


Select a Size

Change View
100 μL
$797.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$797.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

17 kDa

species reactivity

rat, human

concentration

0.5-1 mg/mL

technique(s)

immunoblotting: suitable

accession no.

NM_006763

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BTG2(7832)

General description

B-cell translocation gene 2 (BTG2) belongs to the BTG/TOB gene family. It is also called as nerve growth factor (NGF)-inducible anti-proliferative protein PC3 and NGF-inducible protein TIS21. BTG2 encodes a 158 amino acid protein. This gene is located on the long arm of human chromosome 1.

Immunogen

Synthetic peptide directed towards the middle region of human BTG2

Biochem/physiol Actions

B-cell translocation gene 2 (BTG2) is an anti-proliferative protein. BTG2 modulates transcription regulation mediated by ESR1.
It participates in the modulation of cell cycle transition. It has the ability to repress the migration and invasion of osteosarcoma cells.

Sequence

Synthetic peptide located within the following region: HKMDPIISRVASQIGLSQPQLHQLLPSELTLWVDPYEVSYRIGEDGSICV

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

BTG2: a rising star of tumor suppressors
Mao B, et al.
International journal of oncology, 46(2), 459-464 (2015)
BTG2 inhibits the proliferation and metastasis of osteosarcoma cells by suppressing the PI3K/AKT pathway
Li Yi-Jin, et al.
International Journal of Clinical and Experimental Pathology, 8(10), 12410-12410 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service