Skip to Content
Merck
All Photos(4)

Key Documents

SAB2108151

Sigma-Aldrich

Anti-HNF1B antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-ADTKD3, Anti-FJHN, Anti-HNF-1-beta, Anti-HNF-1B, Anti-HNF1beta, Anti-HNF2, Anti-HPC11, Anti-LF-B3, Anti-LFB3, Anti-MODY5, Anti-RCAD, Anti-T2D, Anti-TCF-2, Anti-TCF2, Anti-VHNF1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$716.00

$716.00


Check Cart for Availability


Select a Size

Change View
100 μL
$716.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$716.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

61kDa

species reactivity

bovine, human, horse, pig

concentration

0.5 mg - 1 mg/mL

technique(s)

immunoblotting: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HNF1B(6928)

Immunogen

Synthetic peptide directed towards the N terminal region of human HNF1B

Biochem/physiol Actions

HNF1B is a member of the transcription factor superfamily. It forms heterodimers with another member of this transcription factor family, HNF1A. Multiple alternatively spliced transcript variants have been described, however the biological validity all of these variants needs to be determined.This gene encodes a member of the transcription factor superfamily. The encoded protein forms heterodimers with another member of this transcription factor family, HNF1A. Multiple alternatively spliced transcript variants have been described, however the biological validity all of these variants needs to be determined. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications. PRIMARYREFSEQ_SPAN PRIMARY_IDENTIFIER PRIMARY_SPAN COMP 1-473 DC340648.1 1-473 474-1994 BC017714.1 441-1961 1995-2544 AC091199.6 29528-30077 2545-2842 AI797324.1 1-298 c

Sequence

Synthetic peptide located within the following region: NFGVKLETLPLSPGSGAEPDTKPVFHTLTNGHAKGRLSGDEGSEDGDDYD

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service