Skip to Content
Merck
All Photos(4)

Documents

SAB2106553

Sigma-Aldrich

Anti-CTSD antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

45 kDa

species reactivity

mouse, human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CTSD(1509)
mouse ... Ctsd(13033)

Immunogen

Synthetic peptide directed towards the C terminal of human CTSD

Biochem/physiol Actions

Ctsd is an acid protease active in intracellular protein breakdown.

Sequence

Synthetic peptide located within the following region: KTICLSGFMGMDIPPPSGPLWILGDVFIGSYYTVFDRDNNRVGFANAVVL

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Madhulika Tripathi et al.
Cell death & disease, 7(12), e2513-e2513 (2016-12-09)
Hyperhomocysteinemia (HHcy) is a well-known risk factor for stroke; however, its underlying molecular mechanism remains unclear. Using both mouse and cell culture models, we have provided evidence that impairment of autophagy has a central role in HHcy-induced cellular injury in
Petya Valcheva et al.
Atherosclerosis, 235(2), 247-255 (2014-06-02)
The inhibition of the renal renin-angiotensin system by the active form of vitamin D contributes to the cardiovascular health benefits of a normal vitamin D status. Local production of angiotensin-II in the vascular wall is a potent mediator of oxidative
Sigrid Regauer et al.
Histology and histopathology, 29(8), 1017-1025 (2014-01-10)
Introduction. Lichen planus (LP) is a chronic cytokine-mediated disease of possible auto-immune etiology. 25% of men have anogenital manifestations. Erosive penile LP causes a scarring phimosis of the foreskin in uncircumcised men. Mast cells as potent immune modulators have been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service