Skip to Content
Merck
All Photos(1)

Key Documents

SAB2105497

Sigma-Aldrich

Anti-FBXL5 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-FBL4, Anti-FBL5, Anti-FLR1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$555.00

$555.00


Check Cart for Availability


Select a Size

Change View
100 μL
$555.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$555.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

63 kDa

species reactivity

guinea pig, rat, rabbit, mouse, human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FBXL5(26234)

Immunogen

Synthetic peptide directed towards the middle region of human FBXL5

Biochem/physiol Actions

FBXL5 is a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. FBXL5 belongs to the Fbls class and, in addition to an F-box, contains several tandem leucine-rich repeats.

Sequence

Synthetic peptide located within the following region: LRTMSSLPESSAMCRKAARTRLPRGKDLIYFGSEKSDQETGRVLLFLSLS

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service