The gene OTUD6B (OTU domain containing 6B) is mapped to human chromosome 8q21. The encoded protein belongs to the OTU (ovarian tumor) family of proteins. OTUD6B is a deubiquitinating enzyme which can be induced by cytokines.
Immunogen
Synthetic peptide directed towards the middle region of human OTUD6B
Biochem/physiol Actions
In B lymphocytes, OTUD6B (OTU domain containing 6B) negatively regulates cell proliferation. In transgenic mouse (TgMMTV-neu) model, OTUD6B autoantibodies were detected prior to the development of breast cancer and might be used for the early human cancer detection.
Sequence
Synthetic peptide located within the following region: EIIQADSPPIIVGEEYSKKPLILVYMRHAYGLGEHYNSVTRLVNIVTENC
Physical form
Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Journal of translational medicine, 12, 121-121 (2014-06-03)
The use of autoantibodies for the early detection of breast cancer has generated much interest as antibodies can be readily assayed in serum when antigen levels are low. Ideally, diagnostic autoantibodies would be identified in individuals who harbored pre-invasive disease/high
Deubiquitinating enzymes (DUBs) are important regulators of cell proliferation. Here we identified a functional deubiquitinating enzyme, ovarian tumor domain-containing 6B (OTUD-6B). Mutation of the conserved Cys residue abolished its deubiquitinating activity in vitro. Otud-6b expression was induced with cytokine stimulation
Epigenetic changes are being increasingly recognized as a prominent feature of cancer. This occurs not only at individual genes, but also over larger chromosomal domains. To investigate this, we set out to identify large chromosomal domains of epigenetic dysregulation in
Questions
Reviews
★★★★★ No rating value
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.