Skip to Content
Merck
All Photos(1)

Key Documents

SAB2103534

Sigma-Aldrich

Anti-MEF2C antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$555.00

$555.00


Estimated to ship on14 May 2025



Select a Size

Change View
100 μL
$555.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$555.00


Estimated to ship on14 May 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

mol wt

51 kDa

species reactivity

mouse, horse, pig, dog, rat, rabbit, human

packaging

pkg of 100 μL buffered aqueous solution
pkg of 50 μg lyophilized powder

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MEF2C(4208)

Immunogen

Synthetic peptide directed towards the middle region of human MEF2C

Sequence

Synthetic peptide located within the following region: PPSALSQLGACTSTHLSQSSNLSLPSTQSLNIKSEPVSPPRDRTTTPSRY

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hongyan Zhu et al.
International journal of molecular medicine, 41(2), 1096-1102 (2017-12-06)
The cofilin 2 (CFL2) and myosin heavy chain (MyHC) genes play a key role in muscle development and myofibrillar formation. The aim of the present study was to investigate the effect of CFL2 on genes involved in fiber formation and the mechanisms

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service