Skip to Content
Merck
All Photos(1)

Key Documents

SAB2103374

Sigma-Aldrich

Anti-SGMS2, (N-terminal) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-MGC26963, Anti-SMS2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$740.00

$740.00


Check Cart for Availability


Select a Size

Change View
100 μL
$740.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$740.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

42 kDa

species reactivity

guinea pig, rabbit, mouse, rat, human, horse, bovine, dog

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SGMS2(166929)

Immunogen

Synthetic peptide directed towards the N terminal region of human SGMS2

Biochem/physiol Actions

SGMS2 is a bidirectional lipid cholinephosphotransferase capable of converting phosphatidylcholine (PC) and ceramide to sphingomyelin (SM) and diacylglycerol (DAG) and vice versa. Direction is dependent on the relative concentrations of DAG and ceramide as phosphocholine acceptors. SGMS2 directly and specifically recognizes the choline head group on the substrate. SGMS2 also requires two fatty chains on the choline-P donor molecule in order to be recognized efficiently as a substrate. SGMS2 does not function strictly as a SM synthase. SGMS2 is required for cell growth.Sphingomyelin (SM) is a major component of plasma membranes. It is preferentially concentrated in the outer leaflet and has a role in the formation of lipid rafts. SM synthases (EC 2.7.8.27), such as SGMS2, produce SM in the lumen of the Golgi and on the cell surface through the transfer of phosphocholine from phosphatidylcholine onto ceramide, yielding diacylglycerol as a side product (Huitema et al., 2004 [PubMed 14685263]).[supplied by OMIM]. Sequence Note: removed 3 bases from the 5′ end that did not align to the reference genome assembly. PRIMARYREFSEQ_SPAN PRIMARY_IDENTIFIER PRIMARY_SPAN COMP 1-2142 BC041369.2 4-2145

Sequence

Synthetic peptide located within the following region: KFPLEWWKTGIAFIYAVFNLVLTTVMITVVHERVPPKELSPPLPDKFFDY

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Animesh Kar et al.
Science advances, 9(26), eadf2746-eadf2746 (2023-06-30)
Treatment of triple-negative breast cancer (TNBC) is challenging because of its "COLD" tumor immunosuppressive microenvironment (TIME). Here, we present a hydrogel-mediated localized delivery of a combination of docetaxel (DTX) and carboplatin (CPT) (called DTX-CPT-Gel therapy) that ensured enhanced anticancer effect

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service