Skip to Content
Merck
All Photos(1)

Key Documents

SAB2102467

Sigma-Aldrich

Anti-TMEM35 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-FLJ14084, Anti-Transmembrane protein 35

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$716.00

$716.00


Check Cart for Availability


Select a Size

Change View
100 μL
$716.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$716.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

18 kDa

species reactivity

guinea pig, human, rabbit, horse, mouse, bovine, rat

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TMEM35(59353)

General description

Transmembrane protein 35 (TMEM35) is mostly expressed in hypothalamic-pituitary-adrenal (HPA) circuitry and limbic areas, including the hippocampus and amygdala of mice. TMEM35 sis also known as the unknown factor-1(TUF-1). Human TMEM35 consists of 167 amino acids and is located at human chromosome Xq22.

Immunogen

Synthetic peptide directed towards the C terminal region of human TMEM35

Biochem/physiol Actions

Transmembrane protein 35 (TMEM35) is a multi-pass membrane protein. Any alteration in Transmembrane protein 35 (TMEM35) causes long term memory loss as well as impairs behavioral functions. TMEM35 regulates cell cycle progression and thereby modulates osteosarcoma cell growth, migration and invasion, making it a potent drug development target for therapeutics. In zona glomerulosa, the neurite outgrowth after sodium depletion is regulated by TMEM35.

Sequence

Synthetic peptide located within the following region: VFGILLTCRLLIARKPEDRSSEKKPLPGNAEEQPSLYEKAPQGKVKVS

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deletion of novel protein TMEM35 alters stress-related functions and impairs long-term memory in mice
Kennedy, , et al.
American Journal of Physiology. Regulatory, Integrative and Comparative Physiology, 311(1), R166-R178 (2016)
Downregulation of coding transmembrane protein 35 gene inhibits cell proliferation, migration and cell cycle arrest in osteosarcoma cells
Huang, Yi, et al.
Experimental and Therapeutic Medicine, 12(2), 581-588 (2016)
Sodium depletion increases sympathetic neurite outgrowth and expression of a novel TMEM35 gene-derived protein (TUF1) in the rat adrenal zona glomerulosa
Tran, Phu, et al.
Endocrinology, 151(10), 4852-4860 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service