Skip to Content
Merck
All Photos(3)

Key Documents

SAB2102261

Sigma-Aldrich

Anti-SOD2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-IPO-B, Anti-MNSOD, Anti-Superoxide dismutase 2, mitochondrial

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$609.00

$609.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1246


Select a Size

Change View
100 μL
$609.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

$609.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1246

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

mol wt

24 kDa

species reactivity

bovine, sheep, rat, dog, goat, human, guinea pig, rabbit

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SOD2(6648)

Immunogen

Synthetic peptide directed towards the N terminal region of human SOD2

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Sequence

Synthetic peptide located within the following region: MLSRAVCGTSRQLAPVLGYLGSRQKHSLPDLPYDYGALEPHINAQIMQLH

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Angelina Rafaela Debortoli et al.
Journal of molecular endocrinology, 59(2), 171-180 (2017-07-25)
Compared with age-matched men, premenopausal women are largely protected from coronary artery disease, a difference that is lost after menopause. The effects of oestrogens are mediated by the activation of nuclear receptors (ERα and ERβ) and by the G protein-coupled
Nathalie T B Delgado et al.
Frontiers in pharmacology, 7, 522-522 (2017-01-20)
Decline in estrogen levels promotes endothelial dysfunction and, consequently, the most prevalent cardiovascular diseases in menopausal women. The use of natural therapies such as pomegranate can change these results. Pomegranate [Punica granatum L. (Punicaceae)] is widely used as a phytotherapeutic
Maki Yoshikawa et al.
Nutrients, 13(4) (2021-04-04)
Fucoxanthin (Fx) has preventive effect against muscle atrophy and myotube loss in vitro, but it has not yet been examined in vivo. Therefore, we aimed to investigate the effect of Fx on dexamethasone (Dex)-induced muscle atrophy and fat mass in
Xuan Xu et al.
Cardiovascular diagnosis and therapy, 10(3), 453-469 (2020-07-23)
Myocardial mitochondrial dysfunction is the leading cause of chronic heart failure (CHF). Increased reactive oxygen species (ROS) levels, disruption of mitochondrial biogenesis and mitochondrial Ca2+([Ca2+]m) homeostasis and reduction of the mitochondrial membrane potential (ΔΨm) cause myocardial mitochondrial dysfunction. Therefore, treating

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service