Skip to Content
Merck
All Photos(3)

Key Documents

SAB2101515

Sigma-Aldrich

Anti-MST1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-D3F15S2, Anti-DNF15S2, Anti-HGFL, Anti-MSP, Anti-Macrophage stimulating 1 (hepatocyte growth factor-like)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$537.00

$537.00


Estimated to ship on01 June 2025



Select a Size

Change View
100 μL
$537.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

$537.00


Estimated to ship on01 June 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

80 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MST1(4485)

Immunogen

Synthetic peptide directed towards the middle region of human MST1

Biochem/physiol Actions

MST1 belongs to the peptidase S1 family, plasminogen subfamily. It contains 4 kringle domains, 1 PAN domain and 1 peptidase S1 domain. MST1 probably has no proteolytic activity, since crucial characteristic of serine proteases catalytic sites are not conserved.

Sequence

Synthetic peptide located within the following region: SCHMPLTGYEVWLGTLFQNPQHGEPSLQRVPVAKMVCGPSGSQLVLLKLE

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service