Skip to Content
Merck
All Photos(3)

Documents

SAB2101225

Sigma-Aldrich

Anti-KCNK9 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-K2p9.1, Anti-KT3.2, Anti-MGC138268, Anti-MGC138270, Anti-Potassium channel, subfamily K, member 9

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

42 kDa

species reactivity

bovine, horse, rabbit, human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KCNK9(51305)

Immunogen

Synthetic peptide directed towards the N terminal region of human KCNK9

Biochem/physiol Actions

KCNK9 is one of the members of the superfamily of potassium channel proteins containing two pore-forming P domains. This open channel is highly expressed in the cerebellum. It is inhibited by extracellular acidification and arachidonic acid, and strongly inhibited by phorbol 12-myristate 13-acetate.This gene encodes one of the members of the superfamily of potassium channel proteins containing two pore-forming P domains. This open channel is highly expressed in the cerebellum. It is inhibited by extracellular acidification and arachidonic acid, and strongly inhibited by phorbol 12-myristate 13-acetate. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.

Sequence

Synthetic peptide located within the following region: REEEKLKAEEIRIKGKYNISSEDYRQLELVILQSEPHRAGVQWKFAGSFY

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masumi Inoue et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 33(1), 455-468 (2018-07-13)
External acidity induces catecholamine secretion by inhibiting TASK1-like channels in rat adrenal medullary (AM) cells. TASK channels can function as a heteromer or homomer in the TASK subfamily. In this study, we elucidate the molecular identity of TASK1-like channels in
Yotesawee Srisomboon et al.
American journal of physiology. Cell physiology, 314(5), C627-C639 (2018-01-25)
The objective of this study was to determine the molecular identity of ion channels involved in K+ secretion by the mammary epithelium and to examine their regulation by purinoceptor agonists. Apical membrane voltage-clamp experiments were performed on human mammary epithelial
Marie Goutierre et al.
Cell reports, 28(1), 91-103 (2019-07-04)
KCC2 regulates neuronal transmembrane chloride gradients and thereby controls GABA signaling in the brain. KCC2 downregulation is observed in numerous neurological and psychiatric disorders. Paradoxical, excitatory GABA signaling is usually assumed to contribute to abnormal network activity underlying the pathology.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service