Skip to Content
Merck
All Photos(1)

Key Documents

SAB2101047

Sigma-Aldrich

Anti-HMG20A antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-FLJ10739, Anti-HMGX1, Anti-High-mobility group 20A

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$537.00

$537.00


Estimated to ship on03 May 2025



Select a Size

Change View
100 μL
$537.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$537.00


Estimated to ship on03 May 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

40 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HMG20A(10363)

General description

HMG20A (high mobility group 20A) is located on human chromosome 15q24. It is a high mobility group (HMG) domain-containing protein.

Immunogen

Synthetic peptide directed towards the N terminal region of human HMG20A

Biochem/physiol Actions

HMG20A (high mobility group 20A) plays a role in neuronal differentiation as chromatin-associated protein. It overcomes the repressive effects of the neuronal silencer REST and induces the activation of neuronal-specific genes. The protein also acts as an inhibitor of HMG20B. It involved in the recruitment of the histone methyltransferase MLL and consequent increased methylation of histone H3 lysine 4.
HMG20A is essential for SNAI1 (snail family transcriptional repressor 1)-mediated epithelial to mesenchymal transition.

Sequence

Synthetic peptide located within the following region: ENLMTSSTLPPLFADEDGSKESNDLATTGLNHPEVPYSSGATSSTNNPEF

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

HMG20A and HMG20B map to human chromosomes 15q24 and 19p13. 3 and constitute a distinct class of HMG-box genes with ubiquitous expression
Sumoy L, et al.
Cytogenetic and genome research, 88(1-2), 62-67 (2000)
HMG20A is required for SNAI1-mediated epithelial to mesenchymal transition
Rivero S, et al.
Oncogene, 34(41), 5264-5264 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service