Skip to Content
Merck
All Photos(2)

Key Documents

SAB2100974

Sigma-Aldrich

Anti-GRIN2A antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Glutamate receptor, ionotropic, N-methyl D-aspartate 2A, Anti-NMDAR2A, Anti-NR2A

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$653.00

$653.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB04901


Select a Size

Change View
100 μL
$653.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

$653.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB04901

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

163 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GRIN2A(2903)

Immunogen

Synthetic peptide directed towards the middle region of human GRIN2A

Biochem/physiol Actions

N-methyl-D-aspartate (NMDA) receptors are a class of ionotropic glutamate-gated ion channels. These receptors have been shown to be involved in long-term potentiation, an activity-dependent increase in the efficiency of synaptic transmission thought to underlie certain kinds of memory and learning. NMDA receptor channels are heteromers composed of the key receptor subunit NMDAR1 (GRIN1) and 1 or more of the 4 NMDAR2 subunits: NMDAR2A (GRIN2A), NMDAR2B (GRIN2B), NMDAR2C (GRIN2C) and NMDAR2D (GRIN2D).N-methyl-D-aspartate (NMDA) receptors are a class of ionotropic glutamate receptors. NMDA channel has been shown to be involved in long-term potentiation, an activity-dependent increase in the efficiency of synaptic transmission thought to underlie certain kinds of memory and learning. NMDA receptor channels are heteromers composed of the key receptor subunit NMDAR1 (GRIN1) and 1 or more of the 4 NMDAR2 subunits: NMDAR2A (GRIN2A), NMDAR2B (GRIN2B), NMDAR2C (GRIN2C), and NMDAR2D (GRIN2D). Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.

Sequence

Synthetic peptide located within the following region: DKIYTIDGEKEPGFHLDPPQFVENVTLPENVDFPDPYQDPSENFRKGDST

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yang Li et al.
Neuroscience bulletin, 35(4), 661-672 (2019-03-23)
The present study was designed to examine the therapeutic effects of Botulinum neurotoxin A (BoNT/A) on depression-like behaviors in mice and to explore the potential mechanisms. These results revealed that a single facial injection of BoNT/A induced a rapid and
Minxia Zhu et al.
Behavioural brain research, 367, 82-90 (2019-04-01)
The present study aimed to observe the effects of acute exposure to hypobaric hypoxia on learning and memory in adult Sprague-Dawley (SD) rats as well as the changes in N-methyl-D-aspartate receptor (NMDAR) subunits. Learning and memory abilities were evaluated with
Yingrak Boondam et al.
Scientific reports, 9(1), 8404-8404 (2019-06-12)
The herb Centella asiatica has long been considered a memory tonic. A recent review found no strong evidence for improvement of cognitive function, suggesting negative results were due to limitations in dose, standardization and product variation. We used a standardized
Lu Luo et al.
Journal of stroke and cerebrovascular diseases : the official journal of National Stroke Association, 28(3), 672-682 (2018-12-07)
High-intensity interval training (HIIT) improves functional and mental health in the patients with stroke. To investigate the potential mechanisms of HIIT on poststroke depression (PSD). Wistar rats were randomly divided into control, Sham, PSD, moderate intensity continuous training (MICT), and
Jean-Nicolas Audet et al.
Science advances, 4(3), eaao6369-eaao6369 (2018-03-17)
Problem solving and innovation are key components of intelligence. We compare wild-caught individuals from two species that are close relatives of Darwin's finches, the innovative Loxigilla barbadensis, and its most closely related species in Barbados, the conservative Tiaris bicolor. We

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service