Skip to Content
Merck
All Photos(2)

Key Documents

SAB2100154

Sigma-Aldrich

Anti-ARNTL2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Aryl hydrocarbon receptor nuclear translocator-like 2, Anti-BMAL2, Anti-CLIF, Anti-MGC149671, Anti-MGC149672

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$609.00

$609.00


Check Cart for Availability


Select a Size

Change View
100 μL
$609.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$609.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

71 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ARNTL2(56938)

Immunogen

Synthetic peptide directed towards the N terminal region of human ARNTL2

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

ARNTL2-CLOCK heterodimers activate E-box element (3′-CACGTG-5′) transcription. Also, in umbilical vein endothelial cells, ARNTL2 activates SERPINE1 through E-box sites. This transactivation is inhibited by PER2 and CRY1.

Sequence

Synthetic peptide located within the following region: PTAMGSFSSHMTEFPRKRKGSDSDPSQSGIMTEKVVEKLSQNPLTYLLST

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tao Wang et al.
BMC molecular and cell biology, 24(1), 14-14 (2023-04-03)
Recent studies have reported that the circadian transcription factor aryl hydrocarbon receptor nuclear translocator like 2 (ARNTL2) promotes the metastatic progression of lung adenocarcinoma. However, the molecular mechanisms of ARNTL2 in non-small cell lung cancer (NSCLC) cell growth and proliferation
Lenka Polidarová et al.
PloS one, 8(9), e75690-e75690 (2013-10-03)
The mammalian timekeeping system generates circadian oscillations that rhythmically drive various functions in the body, including metabolic processes. In the liver, circadian clocks may respond both to actual feeding conditions and to the metabolic state. The temporal restriction of food

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service