Skip to Content
Merck
All Photos(2)

Key Documents

SAB1413160

Sigma-Aldrich

Monoclonal Anti-NFE2L2 (phospho S40) antibody produced in rabbit

clone IFH-14

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$543.00

$543.00


Check Cart for Availability


Select a Size

Change View
100 μG
$543.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$543.00


Check Cart for Availability

biological source

rabbit

antibody product type

primary antibodies

clone

IFH-14, monoclonal

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

flow cytometry: 1:50
immunofluorescence: 1:50-1:200
immunohistochemistry: 1:50-1:200
immunoprecipitation (IP): 1:50
western blot: 1:1000-1:2000

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer40)

Gene Information

human ... NFE2L2(4780)

Related Categories

Immunogen

A synthetic phosphopeptide corresponding to residues surrounding S40 of human NFE2L2.

Physical form

In PBS, 150 mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service