Skip to Content
Merck
All Photos(2)

Key Documents

SAB1412548

Sigma-Aldrich

ANTI-TLR4 antibody produced in mouse

clone 4B10, purified immunoglobulin, buffered aqueous solution

Synonym(s):

ARMD10, CD284, TLR4, TOLL, hToll

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$546.00

$546.00


Check Cart for Availability


Select a Size

Change View
100 μG
$546.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$546.00


Check Cart for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4B10, monoclonal

form

buffered aqueous solution

mol wt

antigen 34.32 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TLR4(7099)

Related Categories

General description

The protein encoded by this gene is a member of the Toll-like receptor (TLR) family which plays a fundamental role in pathogen recognition and activation of innate immunity. TLRs are highly conserved from Drosophila to humans and share structural and functional similarities. They recognize pathogen-associated molecular patterns (PAMPs) that are expressed on infectious agents, and mediate the production of cytokines necessary for the development of effective immunity. The various TLRs exhibit different patterns of expression. This receptor is most abundantly expressed in placenta, and in myelomonocytic subpopulation of the leukocytes. It has been implicated in signal transduction events induced by lipopolysaccharide (LPS) found in most gram-negative bacteria. Mutations in this gene have been associated with differences in LPS responsiveness. Also, several transcript variants of this gene have been found, but the protein coding potential of most of them is uncertain. (provided by RefSeq)
Toll like receptor 4 (TLR4) is an extracellular pathogen recognition receptor (PRR), encoded by the gene mapped to human chromosome 9q32–33. The encoded protein belongs to the interleukin-1 (IL-1)/toll receptor family and is present on both immune and nonimmune cells.

Immunogen

TLR4 (NP_612564, 214 a.a. ~ 291 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
PMNFIQPGAFKEIRLHKLTLRNNFDSLNVMKTCIQGLAGLEVHRLVLGEFRNEGNLEKFDKSALEGLCNLTIEEFRLA

Biochem/physiol Actions

Toll like receptor 4 (TLR4) is a bacterial lipopolysaccharide (LPS) sensor. It plays a vital role in regulation of innate immunity. Palmitic acid (PA) interacts with TLR4 to stimulate pro-inflammatory cytokine interleukin-1β (IL-1β) secretion in human immune cells. Elevated expression of TLR4 is associated with the lupus nephritis (LN) and chronic cutaneous lupus erythematosus (CLE) pathogenesis. Genetic variations in the gene has been observed in patients with esophageal adenocarcinoma (EAC).

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Toll like receptor 4 and hepatocellular carcinoma; A systematic review.
Sepehri Z
Life Sciences, 179, 80-87 (2017)
Impact of mutations in Toll-like receptor pathway genes on esophageal carcinogenesis.
Fels Elliott DR
PLoS Genetics, 13(5), 1-21 (2017)
The Increased Expression of Toll-Like Receptor 4 in Renal and Skin Lesions in Lupus Erythematosus.
Elloumi N
The Journal of Histochemistry and Cytochemistry, 65(7), 389-398 (2017)
Cutting edge: Toll-like receptor 4 (TLR4)-deficient mice are hyporesponsive to lipopolysaccharide: evidence for TLR4 as the Lps gene product.
Hoshino K
Journal of Immunology, 162(7), 3749-3752 (1999)
Palmitic acid is a toll-like receptor 4 ligand that induces human dendritic cell secretion of IL-1?.
Nicholas DA
PLoS ONE, 12(5), 1-24 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service