Skip to Content
Merck
All Photos(1)

Key Documents

SAB1412234

Sigma-Aldrich

ANTI-GPT antibody produced in mouse

clone M1, purified immunoglobulin, buffered aqueous solution

Synonym(s):

AAT1, ALT1, GPT, GPT1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$482.00

$482.00


Estimated to ship on16 May 2025



Select a Size

Change View
100 μG
$482.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$482.00


Estimated to ship on16 May 2025


biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

M1, monoclonal

form

buffered aqueous solution

mol wt

antigen 80.3 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GPT(2875)

General description

Glutamate-pyruvate transaminase (EC 2.6.1.2), also known as alanine aminotransferase, catalyzes the reversible conversion of L-alanine and alpha-ketoglutarate to L-glutamate and pyruvate. It has 2 distinct molecular and genetic forms: one cytoplasmic (soluble) (GPT1) and one mitochondrial (GPT2; MIM 138210). See ALTQTL1 (MIM 612363) and ALTQTL2 (MIM 612364) for information on quantitative trait loci influencing the plasma level of alanine aminotransferase.[supplied by OMIM
The GPT (glutamate pyruvate transaminase 1) gene with 11 exons spanning 2.7kb of genomic DNA, is mapped to human chromosome 8q24.3. The encoded protein consists of 495 amino acids and is identical to the human GPT-1.

Immunogen

GPT (AAH18207.1, 1 a.a. ~ 496 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MASSTGDRSQAVRHGLRAKVLTLDGMNPRVRRVEYAVRGPIVQRALELEQELRQGVKKPFTEVIRANIGDAQAMGQRPITFLRQVLALCVNPDLLSSPNFPDDAKKRAERILQACGGHSLGAYSVSSGIQLIREDVARYIERRDGGIPADPNNVFLSTGASDAIVTVLKLLVAGEGHTRTGVLIPIPQYPLYSATLAELGAVQVDYYLDEERAWALDVAELHRALGQARDHCRPRALCVINPGNPTGQVQTRECIEAVIRFAFEERLFLLADEVYQDNVYAAGSQFHSFKKVLMEMGPPYAGQQELASFHSTSKGYMGECGFRGGYVEVVNMDAAVQQQMLKLMSVRLCPPVPGQALLDLVVSPPAPTDPSFAQFQAEKQAVLAELAAKAKLTEQVFNEAPGISCNPVQGAMYSFPRVQLPPRAVERAQELGLAPDMFFCLRLLEETGICVVPGSGFGQREGTYHFRMTILPPLEKLRLLLEELSRFHAKFTLEYS

Biochem/physiol Actions

Serum GPT (Glutamate pyruvate transaminase 1) acts as a common biomarker in screening chronic liver injury. GPT levels identify the risk of osteoporosis in nonalcoholic fatty liver disease.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Higher Ratio of Serum Alpha-Fetoprotein Could Predict Outcomes in Patients with Hepatitis B Virus-Associated Hepatocellular Carcinoma and Normal Alanine Aminotransferase.
Kim YI
PLoS ONE, 11(6), 1-12 (2016)
The association of liver fat content and serum alanine aminotransferase with bone mineral density in middle-aged and elderly Chinese men and postmenopausal women.
Xia MF
Journal of Translational Medicine, 14:11, 1-20 (2016)
M M Sohocki et al.
Genomics, 40(2), 247-252 (1997-03-01)
Two frequent protein variants of glutamate pyruvate transminase (GPT) (E.C.2.6.1.2) have been used as genetic markers in humans for more than two decades, although chromosomal mapping of the GPT locus in the 1980s produced conflicting results. To resolve this conflict

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service